Gene Transcripts

Gene Symbol
PLEKHF2
Entrez Gene ID
79666
Gene Name
pleckstrin homology and FYVE domain containing 2
Gene Aliases
EAPF, PHAFIN2, ZFYVE18
Chromosome Location
Chr. 8: 95133721 - 95156685 on Build GRCh38
UniGene ID
Hs.29724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024613.3 NP_078889.1 2 649
GenBank mRNA AF434819.1 - 1 325
AK023249.1 - 2 422
AL834473.1 - 2 572
BC011806.2 - 2 549 4335031

View Full Product Details