Gene Transcripts

Gene Symbol
CENPU
Entrez Gene ID
79682
Gene Name
centromere protein U
Gene Aliases
CENP50, CENPU50, KLIP1, MLF1IP, PBIP1
Chromosome Location
Chr. 4: 184694065 - 184734132 on Build GRCh38
UniGene ID
Hs.575032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024629.3 NP_078905.2 11, 12 1050
XM_005263218.4 XP_005263275.2 11, 12 1085
GenBank mRNA AF469667.1 - 11, 12 1053
AF516710.1 - 12, 13 1467
AK027121.1 - 9, 10 860
AK292297.1 - 11, 12 996
BC031520.1 - 6, 7 398
BC107744.1 - 6, 7 495
BC131556.1 - 11, 12 1050 40108113
CR457376.1 - 4, 5 254

View Full Product Details