Gene Transcripts
- Gene Symbol
- MANEA
- Entrez Gene ID
- 79694
- Gene Name
- mannosidase endo-alpha
- Gene Aliases
- ENDO, hEndo
- Chromosome Location
- Chr. 6: 95577497 - 95609452 on Build GRCh38
- UniGene ID
- Hs.734632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024641.3 |
NP_078917.2
|
3, 4 |
842 |
|
XM_005267147.3 |
XP_005267204.1
|
3, 4 |
1235 |
|
GenBank mRNA |
AJ577574.1 |
- |
2, 3 |
707 |
|
AK022900.1 |
- |
2, 3 |
348 |
|
AK291940.1 |
- |
3, 4 |
773 |
|
AY372528.1 |
- |
2, 3 |
662 |
|
BC137014.1 |
- |
2, 3 |
711 |
9021007 |
BC137016.1 |
- |
2, 3 |
711 |
9021009 |
BC146671.1 |
- |
3, 4 |
777 |
40148201 |
BC150199.1 |
- |
3, 4 |
777 |
40148811 |
BX537398.1 |
- |
3, 4 |
773 |
|
BX640869.1 |
- |
3, 4 |
803 |
|
CR936827.1 |
- |
3, 4 |
777 |
|
View Full Product Details