Gene Transcripts

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
Chromosome Location
Chr. X: 45148373 - 45200901 on Build GRCh38
UniGene ID
Hs.98321
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024689.2 NP_078965.2 2 383
NM_176819.3 NP_789789.2 2 383
XM_005272670.1 XP_005272727.1 2 358
XM_006724559.1 XP_006724622.1 2 358
GenBank mRNA AK024165.1 - 2 383
AY358764.1 - 2 341
BC035375.1 - 2 319
BC137528.1 - 2 371
BC137529.1 - 2 371
BP231945.1 - 2 334

View Full Product Details