Gene Transcripts

Gene Symbol
SEH1L
Entrez Gene ID
81929
Gene Name
SEH1 like nucleoporin
Gene Aliases
SEC13L, SEH1A, SEH1B, Seh1
Chromosome Location
Chr. 18: 12947984 - 12987665 on Build GRCh38
UniGene ID
Hs.301048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013437.1 NP_001013455.1 4 550
NM_031216.3 NP_112493.2 4 550
XM_005258152.4 XP_005258209.1 3 393
XM_011525742.2 XP_011524044.1 4 546
XM_011525743.2 XP_011524045.1 3 393
XM_011525744.1 XP_011524046.1 4 546
XM_011525745.2 XP_011524047.1 3 314
XM_011525746.2 XP_011524048.1 4 656
XM_017026025.1 XP_016881514.1 3 399
XM_017026026.1 XP_016881515.1 3 470
XM_017026027.1 XP_016881516.1 4 720
GenBank mRNA AF136976.1 - 4 518
AF255625.1 - 4 454
AF431970.1 - 4 454
AF514996.1 - 4 414
AK056940.1 - 4 513
AK291226.1 - 4 513
AK314403.1 - 4 524
BC012430.1 - 4 504 3865994
CB990796.1 - 577

View Full Product Details