Gene Transcripts

Gene Symbol
WDR87
Entrez Gene ID
83889
Gene Name
WD repeat domain 87
Gene Aliases
NYD-SP11
Chromosome Location
Chr. 19: 37884577 - 37906706 on Build GRCh38
UniGene ID
Hs.664194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291088.1 NP_001278017.1 5 3540
NM_031951.4 NP_114157.4 5 3423
XM_011527360.2 XP_011525662.1 5 3367
GenBank mRNA AK128826.1 - 5 3394
AK302913.1 - 5 3432
AY026504.2 - 5 3451

View Full Product Details