Gene Transcripts

Gene Symbol
KRTAP9-3
Entrez Gene ID
83900
Gene Name
keratin associated protein 9-3
Gene Aliases
KAP9.3, KRTAP9.3
Chromosome Location
Chr. 17: 41232463 - 41233454 on Build GRCh38
UniGene ID
Hs.307012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031962.2 NP_114168.1 1 767
GenBank mRNA AJ406947.1 - 1 767

View Full Product Details