Gene Transcripts
- Gene Symbol
- PHF6
- Entrez Gene ID
- 84295
- Gene Name
- PHD finger protein 6
- Gene Aliases
- BFLS, BORJ, CENP-31
- Chromosome Location
- Chr. X: 134373312 - 134428792 on Build GRCh38
- UniGene ID
- Hs.356501
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001015877.1 |
NP_001015877.1
|
4, 5 |
574 |
|
NM_032335.3 |
NP_115711.2
|
4, 5 |
574 |
|
NM_032458.2 |
NP_115834.1
|
4, 5 |
574 |
|
GenBank mRNA |
AB058726.1 |
- |
4, 5 |
460 |
|
AK290095.1 |
- |
4, 5 |
545 |
|
AK303369.1 |
- |
3, 4 |
437 |
|
AL832725.1 |
- |
4, 5 |
567 |
|
AY157622.1 |
- |
4, 5 |
574 |
|
CD685336.1 |
- |
4, 5 |
582 |
|
DR000282.1 |
- |
4, 5 |
609 |
|
View Full Product Details