Gene Transcripts

Gene Symbol
NTNG2
Entrez Gene ID
84628
Gene Name
netrin G2
Gene Aliases
LHLL9381, Lmnt2, NTNG1, bA479K20.1
Chromosome Location
Chr. 9: 132161234 - 132244534 on Build GRCh38
UniGene ID
Hs.163642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032536.2 NP_115925.2 6, 7 2006
XM_006717304.3 XP_006717367.1 9, 10 2129
XM_011519094.2 XP_011517396.1 11, 12 2345
XM_011519096.2 XP_011517398.1 11, 12 2742
XM_011519097.2 XP_011517399.1 10, 11 2263
XM_011519098.2 XP_011517400.1 11, 12 2295
XM_011519099.2 XP_011517401.1 10, 11 2266
XM_011519100.2 XP_011517402.1 9, 10 2195
XM_011519102.2 XP_011517404.1 10, 11 2210
XM_011519103.2 XP_011517405.1 9, 10 2102
XM_011519104.2 XP_011517406.1 9, 10 2326
XM_011519105.2 XP_011517407.1 9, 10 2192
XM_011519106.2 XP_011517408.1 10, 11 2276
XM_011519107.2 XP_011517409.1 8, 9 2027
XM_011519112.2 XP_011517414.1 4, 5 1314
XM_011519113.2 XP_011517415.1 3, 4 1245
XM_017015212.1 XP_016870701.1 12, 13 2449
XM_017015213.1 XP_016870702.1 10, 11 2652
XM_017015214.1 XP_016870703.1 9, 10 2174
GenBank mRNA AB058760.2 - 6, 7 2006
AF131842.1 - 4, 5 420
BC013770.1 - 5, 6 1315 4344617

View Full Product Details