Gene Transcripts
- Gene Symbol
- HIST1H2BK
- Entrez Gene ID
- 85236
- Gene Name
- histone cluster 1, H2bk
- Gene Aliases
- H2B/S, H2BFAiii, H2BFT, H2BK
- Chromosome Location
- Chr. 6: 27138293 - 27146858 on Build GRCh38
- UniGene ID
- Hs.437275
- Species
- Homo sapiens
- Gene Symbol
- H2BFS
- Entrez Gene ID
- 54145
- Gene Name
- H2B histone family member S (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 21: 43565239 - 43565617 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC126339.1 |
- |
1 |
102 |
|
BC126369.1 |
- |
1 |
102 |
|
- Gene Symbol
- LOC102724334
- Entrez Gene ID
- 102724334
- Gene Name
- histone H2B type F-S-like
- Gene Aliases
- Chromosome Location
- Chr. 21: 5972923 - 5973357 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details