Gene Transcripts
- Gene Symbol
- SHANK3
- Entrez Gene ID
- 85358
- Gene Name
- SH3 and multiple ankyrin repeat domains 3
- Gene Aliases
- DEL22q13.3, PROSAP2, PSAP2, SCZD15, SPANK-2
- Chromosome Location
- Chr. 22: 50674642 - 50733212 on Build GRCh38
- UniGene ID
- Hs.149035
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033517.1 |
NP_277052.1
|
9, 10 |
1027 |
|
GenBank mRNA |
AB569469.1 |
- |
9, 10 |
1027 |
|
View Full Product Details