Gene Transcripts

Gene Symbol
MCU
Entrez Gene ID
90550
Gene Name
mitochondrial calcium uniporter
Gene Aliases
C10orf42, CCDC109A
Chromosome Location
Chr. 10: 72692131 - 72887694 on Build GRCh38
UniGene ID
Hs.591366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270679.1 NP_001257608.1 6, 7 827
NM_001270680.1 NP_001257609.1 6, 7 1167
NM_138357.2 NP_612366.1 6, 7 890
NR_073062.1 8, 9 1055
XM_017016882.1 XP_016872371.1 6, 7 1090
GenBank mRNA AK025699.1 - 2, 3 232
AK075439.1 - 6, 7 877
AK128016.1 - 6, 7 1167
AK301475.1 - 8, 9 1036
AK315519.1 - 6, 7 873
BC010682.1 - 6, 7 784
BC034235.1 - 6, 7 890 5296557

View Full Product Details