Gene Transcripts

Gene Symbol
BOD1
Entrez Gene ID
91272
Gene Name
biorientation of chromosomes in cell division 1
Gene Aliases
FAM44B
Chromosome Location
Chr. 5: 173607145 - 173617077 on Build GRCh38
UniGene ID
Hs.425091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138369.2 NP_612378.1 3 641
GenBank mRNA AK301982.1 - 3 629
AY303777.1 - 3 414
BC007436.2 - 3 631 2961368

View Full Product Details