Gene Transcripts

Gene Symbol
MRRF
Entrez Gene ID
92399
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
MRFF, MTRRF, RRF
Chromosome Location
Chr. 9: 122264603 - 122323464 on Build GRCh38
UniGene ID
Hs.368011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138777.3 NP_620132.1 3 612
NM_199177.2 NP_954646.1 2 276
XM_005252302.3 XP_005252359.1 4 680
XM_011519183.2 XP_011517485.1 4 935
XM_011519184.2 XP_011517486.1 4 445
XM_011519185.2 XP_011517487.1 4 680
XM_011519186.1 XP_011517488.1 3 360
XM_011519187.2 XP_011517489.1 2 400
XM_017015286.1 XP_016870775.1 4 680
XM_017015287.1 XP_016870776.1 3 360
GenBank mRNA AK291603.1 - 3 302
AK291809.1 - 3 341
AK292790.1 - 3 341
AK298046.1 - 3 331
AK310974.1 - 2 400
AK316266.1 - 4 629
BC013049.1 - 2 273 3882448
BC053604.1 - 2 274 6008684
BF982227.1 - 347

View Full Product Details