Gene Transcripts

Gene Symbol
LRRC4B
Entrez Gene ID
94030
Gene Name
leucine rich repeat containing 4B
Gene Aliases
HSM, LRIG4
Chromosome Location
Chr. 19: 50516892 - 50568208 on Build GRCh38
UniGene ID
Hs.120873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080457.1 NP_001073926.1 3 2231
XM_005259429.4 XP_005259486.1 3 2410
XM_006723506.3 XP_006723569.1 4 2512
XM_011527520.2 XP_011525822.1 3 3658
GenBank mRNA BC056207.1 - 1 438
BC131897.1 - 4 1328

View Full Product Details