Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052967.1 | NP_443199.1 | 1 | 739 | |
GenBank mRNA | BC069127.1 | - | 1 | 739 | |
BC069345.1 | - | 1 | 739 | 7262308 | |
BC095511.1 | - | 1 | 804 | 30706603 | |
BC101176.2 | - | 1 | 739 | 40016384 | |
BC101177.1 | - | 1 | 739 | 40016387 |