Gene Transcripts
- Gene Symbol
- HSPA12B
- Entrez Gene ID
- 116835
- Gene Name
- heat shock protein family A (Hsp70) member 12B
- Gene Aliases
- C20orf60
- Chromosome Location
- Chr. 20: 3732670 - 3753111 on Build GRCh38
- UniGene ID
- Hs.516854
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197327.1 |
NP_001184256.1
|
3, 4 |
275 |
|
NM_052970.4 |
NP_443202.3
|
3, 4 |
275 |
|
XM_017027632.1 |
XP_016883121.1
|
3, 4 |
190 |
|
XM_017027633.1 |
XP_016883122.1
|
3, 4 |
174 |
|
XM_017027634.1 |
XP_016883123.1
|
3, 4 |
174 |
|
XM_017027635.1 |
XP_016883124.1
|
3, 4 |
218 |
|
GenBank mRNA |
AK056712.1 |
- |
3, 4 |
212 |
|
BC110881.1 |
- |
3, 4 |
224 |
5725594 |
BC117284.1 |
- |
3, 4 |
173 |
40125835 |
BC126263.1 |
- |
3, 4 |
173 |
8991979 |
BC143932.1 |
- |
3, 4 |
173 |
9052450 |
CB990660.1 |
- |
3, 4 |
305 |
|
View Full Product Details