Gene Transcripts

Gene Symbol
TMIGD2
Entrez Gene ID
126259
Gene Name
transmembrane and immunoglobulin domain containing 2
Gene Aliases
CD28H, IGPR-1, IGPR1
Chromosome Location
Chr. 19: 4292227 - 4302439 on Build GRCh38
UniGene ID
Hs.263928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169126.1 NP_001162597.1 3 474
NM_001308232.1 NP_001295161.1 2 114
NM_144615.2 NP_653216.2 3 474
XM_017026284.1 XP_016881773.1 3 482
GenBank mRNA AY358964.1 - 3 436
BC015655.1 - 3 474 4811204
HQ452865.1 - 2 68

View Full Product Details