Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136504.1 | NP_001129976.1 | 5 | 776 | |
NM_177402.4 | NP_796376.2 | 5 | 766 | ||
XM_011509192.2 | XP_011507494.1 | 5 | 796 | ||
XM_017000309.1 | XP_016855798.1 | 4 | 2542 | ||
XM_017000310.1 | XP_016855799.1 | 4 | 2533 | ||
XM_017000311.1 | XP_016855800.1 | 5 | 785 | ||
XM_017000312.1 | XP_016855801.1 | 5 | 1450 | ||
XM_017000313.1 | XP_016855802.1 | 5 | 1440 | ||
GenBank mRNA | AK090672.1 | - | 5 | 760 | |
AK094430.1 | - | 5 | 776 | ||
BC100814.1 | - | 5 | 631 | 40000996 | |
BC100815.1 | - | 5 | 631 | 40000997 | |
BC100816.1 | - | 3 | 278 | ||
BC100817.1 | - | 5 | 631 | 40001000 |