Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017928.3 | NP_001017928.1 | 4, 5 | 392 | |
NM_001308326.1 | NP_001295255.1 | 4, 5 | 418 | ||
XM_011512410.2 | XP_011510712.1 | 4, 5 | 370 | ||
GenBank mRNA | BC054051.1 | - | 4, 5 | 383 | |
BC062725.1 | - | 4, 5 | 392 | 6495960 | |
BC109378.1 | - | 4, 5 | 376 | ||
CD683826.1 | - | 4, 5 | 414 | ||
DA025111.1 | - | 4, 5 | 414 |