Gene Transcripts

Gene Symbol
MROH2B
Entrez Gene ID
133558
Gene Name
maestro heat like repeat family member 2B
Gene Aliases
HEATR7B2
Chromosome Location
Chr. 5: 40998020 - 41071342 on Build GRCh38
UniGene ID
Hs.97714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173489.4 NP_775760.3 34 4221
XM_011513952.1 XP_011512254.1 34 4207
XM_011513953.1 XP_011512255.1 33 4145
GenBank mRNA AF432068.1 - 4 586
AK097562.1 - 10 1267
AK126005.1 - 19 2090
AK128183.1 - 24 2687
AL832763.1 - 29 3146
BC044937.1 - 22 2693
BC052962.1 - 34 4182 4838071
CR749346.1 - 29 3122

View Full Product Details