Gene Transcripts

Gene Symbol
SPANXN3
Entrez Gene ID
139067
Gene Name
SPANX family member N3
Gene Aliases
CT11.8, SPANX-N3
Chromosome Location
Chr. X: 143508735 - 143517475 on Build GRCh38
UniGene ID
Hs.447320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009609.3 NP_001009609.2 1, 2 169
GenBank mRNA BC062750.1 - 173
DQ336128.1 - 1, 2 85
DQ336129.1 - 1, 2 85

View Full Product Details