Gene Transcripts

Gene Symbol
ZNF362
Entrez Gene ID
149076
Gene Name
zinc finger protein 362
Gene Aliases
RN, lin-29
Chromosome Location
Chr. 1: 33256307 - 33300719 on Build GRCh38
UniGene ID
Hs.743358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152493.2 NP_689706.2 5, 6 853
XM_005270507.2 XP_005270564.1 4, 5 993
XM_005270508.4 XP_005270565.1 5, 6 1650
XM_017000415.1 XP_016855904.1 4, 5 1565
GenBank mRNA AF318356.1 - 4, 5 703
BC104779.1 - 4, 5 775 8143782
BC112113.1 - 4, 5 775 8327581

View Full Product Details