Gene Transcripts

Gene Symbol
ELAVL4
Entrez Gene ID
1996
Gene Name
ELAV like neuron-specific RNA binding protein 4
Gene Aliases
HUD, PNEM
Chromosome Location
Chr. 1: 50048014 - 50203786 on Build GRCh38
UniGene ID
Hs.213050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144774.2 NP_001138246.1 3 533
NM_001144775.2 NP_001138247.2 3 383
NM_001144776.2 NP_001138248.1 3 345
NM_001144777.2 NP_001138249.1 3 423
NM_001294348.1 NP_001281277.1 3 603
NM_001324208.1 NP_001311137.1 3 423
NM_001324209.1 NP_001311138.1 4 620
NM_001324212.1 NP_001311141.1 4 620
NM_001324213.1 NP_001311142.1 3 603
NM_001324214.1 NP_001311143.1 4 690
NM_001324215.1 NP_001311144.1 3 603
NM_001324216.1 NP_001311145.1 3 533
NM_001324217.1 NP_001311146.1 3 603
NM_021952.4 NP_068771.2 3 533
NR_136725.1 4 690
XM_006710411.3 XP_006710474.1 3 515
XM_011540889.2 XP_011539191.1 3 398
XM_011540890.2 XP_011539192.1 3 397
XM_011540893.2 XP_011539195.1 3 345
XM_011540894.2 XP_011539196.1 4 687
XM_011540895.2 XP_011539197.1 4 591
XM_017000538.1 XP_016856027.1 3 517
XM_017000539.1 XP_016856028.1 3 1144
XM_017000540.1 XP_016856029.1 3 401
XM_017000541.1 XP_016856030.1 3 533
XM_017000542.1 XP_016856031.1 3 382
GenBank mRNA AK297338.1 - 3 423
AK298817.1 - 3 412
AK298902.1 - 3 603
AK309653.1 - 3 378
AY033995.1 - 3 345
AY033996.1 - 3 356
AY033997.1 - 3 520
AY033998.1 - 3 487
BC036071.1 - 3 491 5286347
BM509527.1 - 3 390
DC351092.1 - 3 423
M62843.1 - 3 357

View Full Product Details