Gene Transcripts

Gene Symbol
PPTC7
Entrez Gene ID
160760
Gene Name
PTC7 protein phosphatase homolog
Gene Aliases
TA-PP2C, TAPP2C
Chromosome Location
Chr. 12: 110534432 - 110583259 on Build GRCh38
UniGene ID
Hs.13854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139283.1 NP_644812.1 2 558
GenBank mRNA AF385435.1 - 2 558
AK124744.1 - 3 402
AY357944.1 - 2 327
BC111551.1 - 3 360 40009728

View Full Product Details