Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286434.1 | NP_001273363.1 | 9 | 932 | |
NM_153356.2 | NP_699187.1 | 10 | 1040 | ||
XM_006720409.3 | XP_006720472.1 | 10 | 1070 | ||
XM_006720410.3 | XP_006720473.1 | 9 | 962 | ||
XM_006720411.3 | XP_006720474.1 | 8 | 858 | ||
XM_011521281.2 | XP_011519583.1 | 10 | 1069 | ||
XM_011521282.2 | XP_011519584.1 | 10 | 1069 | ||
XM_011521283.2 | XP_011519585.1 | 10 | 1069 | ||
XM_011521284.2 | XP_011519586.1 | 9 | 849 | ||
GenBank mRNA | AB449907.1 | - | 9 | 808 | |
BC033516.1 | - | 10 | 999 | 5170475 |