Gene Transcripts

Gene Symbol
PLAC4
Entrez Gene ID
191585
Gene Name
placenta specific 4
Gene Aliases
C21orf115, PRED78
Chromosome Location
Chr. 21: 41175231 - 41185239 on Build GRCh38
UniGene ID
Hs.472492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182832.2 NP_878252.2 1 6948
GenBank mRNA AK027868.1 - 1 4610
AK092431.1 - 2 1601

View Full Product Details