Gene Transcripts

Gene Symbol
PLCXD2
Entrez Gene ID
257068
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 2
Gene Aliases
Chromosome Location
Chr. 3: 111674676 - 111846447 on Build GRCh38
UniGene ID
Hs.477114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185106.1 NP_001172035.1 2 872
NM_153268.3 NP_695000.1 2 872
GenBank mRNA AK056141.1 - 2 872
BC121155.1 - 2 626 40121377
BC121156.1 - 2 626 40121378

View Full Product Details