Gene Transcripts

Gene Symbol
SLC6A19
Entrez Gene ID
340024
Gene Name
solute carrier family 6 member 19
Gene Aliases
B0AT1, HND
Chromosome Location
Chr. 5: 1201595 - 1225117 on Build GRCh38
UniGene ID
Hs.481478
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003841.2 NP_001003841.1 3 429
GenBank mRNA AK290811.1 - 3 429
AY591756.1 - 3 373
AY596807.1 - 3 373

View Full Product Details