Gene Transcripts

Gene Symbol
TCEAL5
Entrez Gene ID
340543
Gene Name
transcription elongation factor A like 5
Gene Aliases
WEX4
Chromosome Location
Chr. X: 103273690 - 103276869 on Build GRCh38
UniGene ID
Hs.403462
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012979.2 NP_001012997.1 3 997
GenBank mRNA AF086094.1 - 1 99

View Full Product Details