Gene Transcripts

Gene Symbol
OSTN
Entrez Gene ID
344901
Gene Name
osteocrin
Gene Aliases
MUSCLIN
Chromosome Location
Chr. 3: 191199060 - 191265601 on Build GRCh38
UniGene ID
Hs.526794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198184.1 NP_937827.1 3 329
XM_005247428.4 XP_005247485.1 4 577
XM_017006303.1 XP_016861792.1 4 488
GenBank mRNA AY398681.1 - 3 329
AY573933.1 - 3 329
BC128106.1 - 3 329 40112103
BC128107.1 - 3 329 40112104

View Full Product Details