Gene Transcripts
- Gene Symbol
- TBC1D10C
- Entrez Gene ID
- 374403
- Gene Name
- TBC1 domain family member 10C
- Gene Aliases
- CARABIN, EPI64C
- Chromosome Location
- Chr. 11: 67403718 - 67410090 on Build GRCh38
- UniGene ID
- Hs.534648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256508.1 |
NP_001243437.1
|
3, 4 |
286 |
|
NM_198517.3 |
NP_940919.1
|
3, 4 |
303 |
|
NR_046266.1 |
|
2, 3 |
473 |
|
XM_006718538.3 |
XP_006718601.1
|
3, 4 |
562 |
|
XM_006718539.3 |
XP_006718602.1
|
3, 4 |
545 |
|
XM_006718540.3 |
XP_006718603.1
|
3, 4 |
475 |
|
XM_006718541.3 |
XP_006718604.1
|
3, 4 |
570 |
|
XM_006718542.3 |
XP_006718605.1
|
3, 4 |
553 |
|
XM_006718543.2 |
XP_006718606.1
|
3, 4 |
287 |
|
XM_006718544.3 |
XP_006718607.1
|
3, 4 |
481 |
|
XM_011545001.2 |
XP_011543303.1
|
2, 3 |
508 |
|
XM_011545002.2 |
XP_011543304.1
|
2, 3 |
523 |
|
GenBank mRNA |
BC022202.1 |
- |
2, 3 |
247 |
|
BC036873.1 |
- |
2, 3 |
258 |
5224063 |
BC062999.1 |
- |
3, 4 |
275 |
5176854 |
CR979868.1 |
- |
3, 4 |
303 |
|
View Full Product Details