Gene Transcripts

Gene Symbol
OR52K1
Entrez Gene ID
390036
Gene Name
olfactory receptor family 52 subfamily K member 1
Gene Aliases
OR11-8
Chromosome Location
Chr. 11: 4488879 - 4489908 on Build GRCh38
UniGene ID
Hs.553726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005171.2 NP_001005171.2 1 924
GenBank mRNA BC137056.1 - 1 924
BC137059.1 - 1 924 9021056

View Full Product Details