Gene Transcripts

Gene Symbol
OR51I2
Entrez Gene ID
390064
Gene Name
olfactory receptor family 51 subfamily I member 2
Gene Aliases
OR11-38
Chromosome Location
Chr. 11: 5453408 - 5454477 on Build GRCh38
UniGene ID
Hs.553733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004754.2 NP_001004754.1 1 674
GenBank mRNA BC140730.1 - 1 674

View Full Product Details