Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000255.3 | NP_000246.2 | 2 | 481 | |
XM_005249143.3 | XP_005249200.1 | 2 | 386 | ||
GenBank mRNA | AK292568.1 | - | 2 | 457 | |
AK312611.1 | - | 2 | 456 | ||
BC016282.1 | - | 2 | 344 | 3908548 | |
BT007434.1 | - | 1 | 216 | ||
BX647789.1 | - | 2 | 306 | ||
DA777441.1 | - | 2 | 488 | ||
DB290266.1 | - | 2 | 471 | ||
KF030882.1 | - | 1 | 216 | ||
KU178201.1 | - | 1 | 216 | ||
KU178202.1 | - | 1 | 216 | ||
M65131.1 | - | 2 | 292 |