Gene Transcripts

Gene Symbol
MANBA
Entrez Gene ID
4126
Gene Name
mannosidase beta
Gene Aliases
MANB1
Chromosome Location
Chr. 4: 102631486 - 102760998 on Build GRCh38
UniGene ID
Hs.480415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005908.3 NP_005899.3 13 1936
XM_011531966.2 XP_011530268.1 8 1071
XM_017008203.1 XP_016863692.1 14 1922
XM_017008204.1 XP_016863693.1 12 1767
XM_017008205.1 XP_016863694.1 4 2732
GenBank mRNA AB209846.1 - 13 6071
AK291598.1 - 13 1924
AK300015.1 - 12 1706
AK312827.1 - 13 1889
BC015743.1 - 13 1926 4905535
EU009130.1 - 13 1836
U60337.1 - 13 1923

View Full Product Details