Gene Transcripts

Gene Symbol
OR2AK2
Entrez Gene ID
391191
Gene Name
olfactory receptor family 2 subfamily AK member 2
Gene Aliases
OR1-47, OR2AK1P
Chromosome Location
Chr. 1: 247965332 - 247966339 on Build GRCh38
UniGene ID
Hs.690413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004491.1 NP_001004491.1 1 340
GenBank mRNA BC136816.1 - 1 439

View Full Product Details