Gene Transcripts

Gene Symbol
MFN2
Entrez Gene ID
9927
Gene Name
mitofusin 2
Gene Aliases
CMT2A, CMT2A2, CPRP1, HMSN6A, HSG, MARF
Chromosome Location
Chr. 1: 11980181 - 12013515 on Build GRCh38
UniGene ID
Hs.376681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127660.1 NP_001121132.1 2, 3 480
NM_014874.3 NP_055689.1 3, 4 625
XM_005263543.3 XP_005263600.1 4, 5 638
XM_005263545.3 XP_005263602.1 3, 4 488
XM_005263547.3 XP_005263604.1 3, 4 520
XM_005263548.3 XP_005263605.1 4, 5 655
GenBank mRNA AF036536.2 - 3, 4 494
AK289828.1 - 2, 3 425
AK295890.1 - 3, 4 360
AY028429.1 - 2, 3 198
BC017061.1 - 3, 4 355 3901235
BU178912.1 - 3, 4 420
D86987.1 - 3, 4 494

View Full Product Details