Gene Transcripts

Gene Symbol
PLEKHM1
Entrez Gene ID
9842
Gene Name
pleckstrin homology and RUN domain containing M1
Gene Aliases
AP162, B2, OPTB6
Chromosome Location
Chr. 17: 45434262 - 45490780 on Build GRCh38
UniGene ID
Hs.514242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014798.2 NP_055613.1 4, 5 1100
NR_027774.1 3, 4 852
XM_006722201.3 XP_006722264.1 2, 3 881
XM_011525523.2 XP_011523825.1 4, 5 1071
XM_011525524.1 XP_011523826.1 4, 5 1250
XM_011525525.1 XP_011523827.1 3, 4 858
XM_011525526.1 XP_011523828.1 5, 6 894
XM_017025451.1 XP_016880940.1 4, 5 1048
XM_017025452.1 XP_016880941.1 3, 4 803
XM_017025453.1 XP_016880942.1 5, 6 1257
GenBank mRNA AB002354.2 - 4, 5 1053
AJ002220.1 - 2, 3 688
AK299116.1 - 3, 4 800
AK299467.1 - 3, 4 854
BC064361.1 - 4, 5 1064 5535600

Gene Symbol
PLEKHM1P1
Entrez Gene ID
440456
Gene Name
pleckstrin homology and RUN domain containing M1 pseudogene 1
Gene Aliases
PLEKHM1P
Chromosome Location
Chr. 17: 64784841 - 64837184 on Build GRCh38
UniGene ID
Hs.646585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK308180.1 - 5, 6 863

View Full Product Details