Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000436.3 | NP_000427.1 | 17 | 2213 | |
XM_017009494.1 | XP_016864983.1 | 16 | 2004 | ||
GenBank mRNA | AK125115.1 | - | 1 | 2789 | |
AK298352.1 | - | 6 | 779 | ||
AK299668.1 | - | 13 | 1401 | ||
AK315902.1 | - | 6 | 861 | ||
BC009001.1 | - | 17 | 1937 | 4148643 | |
U62961.1 | - | 17 | 1980 |