Gene Transcripts

Gene Symbol
NOTCH3
Entrez Gene ID
4854
Gene Name
notch 3
Gene Aliases
CADASIL, CADASIL1, CASIL, IMF2, LMNS
Chromosome Location
Chr. 19: 15159633 - 15200981 on Build GRCh38
UniGene ID
Hs.8546
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000435.2 NP_000426.2 32 5978
XM_005259924.4 XP_005259981.1 31 5761
GenBank mRNA U97669.1 - 32 5980

View Full Product Details