Gene Transcripts

Gene Symbol
UGT1A5
Entrez Gene ID
54579
Gene Name
UDP glucuronosyltransferase family 1 member A5
Gene Aliases
UDPGT, UDPGT 1-5, UGT1E
Chromosome Location
Chr. 2: 233712992 - 233773299 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019078.1 NP_061951.1 1 364
GenBank mRNA AY435140.1 - 1 364
DQ383514.1 - 1 73

View Full Product Details