Gene Transcripts

Gene Symbol
TIMM8A
Entrez Gene ID
1678
Gene Name
translocase of inner mitochondrial membrane 8 homolog A (yeast)
Gene Aliases
DDP, DDP1, DFN1, MTS, TIM8
Chromosome Location
Chr. X: 101345656 - 101348969 on Build GRCh38
UniGene ID
Hs.447877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145951.1 NP_001139423.1 1 129
NM_004085.3 NP_004076.1 1 378
GenBank mRNA AK312117.1 - 1 126
BC005236.1 - 1 127
BC006994.1 - 1 128 4051368
BC015093.1 - 1 131 3684305
BC070284.1 - 1 111 4281013
BM467820.1 - 1 129
CN410182.1 - 1 378
U66035.1 - 1 108

View Full Product Details