Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031471.5 | NP_113659.3 | 11 | 1368 | |
NM_178443.2 | NP_848537.1 | 11 | 1380 | ||
XM_011545294.2 | XP_011543596.1 | 11 | 1392 | ||
XM_011545295.2 | XP_011543597.1 | 10 | 1225 | ||
XM_017018398.1 | XP_016873887.1 | 11 | 1376 | ||
XM_017018399.1 | XP_016873888.1 | 10 | 1228 | ||
GenBank mRNA | AK093719.1 | - | 11 | 1313 | |
AY093951.1 | - | 11 | 1314 | ||
AY093952.1 | - | 11 | 1326 | ||
BC004347.1 | - | 11 | 1312 | 3634032 | |
BC013366.2 | - | 11 | 1312 | 3677274 | |
BC015584.2 | - | 11 | 1307 | 4637808 |