Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000295.4 | NP_000286.3 | 2 | 808 | |
NM_001002235.2 | NP_001002235.1 | 2 | 787 | ||
NM_001002236.2 | NP_001002236.1 | 4 | 1101 | ||
NM_001127700.1 | NP_001121172.1 | 2 | 824 | ||
NM_001127701.1 | NP_001121173.1 | 4 | 1120 | ||
NM_001127702.1 | NP_001121174.1 | 3 | 928 | ||
NM_001127703.1 | NP_001121175.1 | 4 | 1083 | ||
NM_001127704.1 | NP_001121176.1 | 4 | 1080 | ||
NM_001127705.1 | NP_001121177.1 | 4 | 1098 | ||
NM_001127706.1 | NP_001121178.1 | 3 | 891 | ||
NM_001127707.1 | NP_001121179.1 | 3 | 888 | ||
XM_017021370.1 | XP_016876859.1 | 3 | 988 | ||
GenBank mRNA | AK026174.1 | - | 2 | 590 | |
AK315637.1 | - | 3 | 844 | ||
BC011991.1 | - | 2 | 779 | 3859644 | |
BC015642.2 | - | 2 | 574 | 4644658 | |
BC070163.1 | - | 3 | 514 | ||
BT019455.1 | - | 1 | 547 | ||
BX161449.1 | - | 2 | 714 | ||
BX247968.1 | - | 4 | 1028 | ||
BX248002.1 | - | 1 | 556 | ||
BX248257.1 | - | 2 | 581 | ||
DQ682455.1 | - | 1 | 547 | ||
GU727620.1 | - | 1 | 548 | ||
K01396.1 | - | 1 | 566 | ||
KU178257.1 | - | 1 | 547 | ||
M11465.1 | - | 1 | 558 | ||
X01683.1 | - | 1 | 585 | ||
X17122.1 | - | 1 | 126 |