Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001257180.1 | NP_001244109.1 | 3 | 1073 | |
NM_001257181.1 | NP_001244110.1 | 3 | 704 | ||
NM_006749.4 | NP_006740.1 | 3 | 1036 | ||
XM_005273613.3 | XP_005273670.1 | 3 | 811 | ||
XM_005273615.3 | XP_005273672.1 | 3 | 811 | ||
XM_006716391.3 | XP_006716454.1 | 4 | 4291 | ||
XM_017013748.1 | XP_016869237.1 | 3 | 927 | ||
XM_017013750.1 | XP_016869239.1 | 4 | 946 | ||
XM_017013751.1 | XP_016869240.1 | 4 | 820 | ||
XM_017013752.1 | XP_016869241.1 | 4 | 1062 | ||
GenBank mRNA | AK291202.1 | - | 3 | 1073 | |
AK314774.1 | - | 3 | 705 | ||
BC028600.2 | - | 3 | 748 | 4820601 | |
L20852.1 | - | 2 | 622 |