Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198586.2 | NP_940988.2 | 1 | 1071 | |
GenBank mRNA | AY324850.1 | - | 1 | 1071 | |
BC103888.1 | - | 1 | 1071 | 40004980 | |
BC103889.1 | - | 1 | 1071 | 40004983 | |
BC103890.1 | - | 1 | 1071 | 40004985 |