Gene Transcripts

Gene Symbol
KLHL31
Entrez Gene ID
401265
Gene Name
kelch like family member 31
Gene Aliases
BKLHD6, KBTBD1, KLHL, bA345L23.2
Chromosome Location
Chr. 6: 53647901 - 53665854 on Build GRCh38
UniGene ID
Hs.131064
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003760.4 NP_001003760.2 2 1183
XM_005249107.4 XP_005249164.1 2 1378
XM_017010858.1 XP_016866347.1 2 2293
GenBank mRNA BC137267.1 - 1 1068
BC137269.1 - 1 1068 9021266
EF633513.1 - 2 1183

View Full Product Details