Gene Transcripts

Gene Symbol
LRRD1
Entrez Gene ID
401387
Gene Name
leucine rich repeats and death domain containing 1
Gene Aliases
Chromosome Location
Chr. 7: 92144884 - 92165276 on Build GRCh38
UniGene ID
Hs.671729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161528.1 NP_001155000.1 2 2154
GenBank mRNA BC026112.1 - 2 690
BC144674.1 - 2 249 9053208
BC144678.1 - 2 272 9053212

View Full Product Details