Gene Transcripts

Gene Symbol
OR4F21
Entrez Gene ID
441308
Gene Name
olfactory receptor family 4 subfamily F member 21
Gene Aliases
OR4F21P
Chromosome Location
Chr. 8: 166086 - 167024 on Build GRCh38
UniGene ID
Hs.690459
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005504.1 NP_001005504.1 1 121

View Full Product Details